Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics™ SARS-CoV-2 Synthetic RNA (N Gene Targets)
SDP

Catalog No. 23004149 Shop All Microbiologics Products
Change view
Click to view available options
Quantity:
1 Vial
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-004-149 1 Vial
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-004-149 Supplier Microbiologics™ Supplier No. HE0060S
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Synthetic RNA molecular standard intended for use as a positive control in molecular applications. Kit includes 1 vial of dried synthetic RNA (up to 100 reactions), 1 vial of molecular standard water for rehydration, and a Certificate of Analysis.

  • Strain Characteristics: The SARS-CoV-2 synthetic RNA material is a 1044-nucleotide portion of the SARS-CoV-2 N (nucleocapsid) gene. It contains three markers N1, N2 and N3 that are commonly used in diagnostic assays
  • IVD
  • Dried RNA Preserved with Biomatrica RNAstable™

Specifications

Biosafety Level 1
Content And Storage 2°C to 25°C
Type Pellet
For Use With (Application) Diagnostic Assays
Packaging Plastic Box
Quantity 1 Vial
Source United States
Form Pellet
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.