Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics™ SARS-CoV-2 Synthetic RNA Pool (N/E/RdRp/S Gene Targets)
SDP

Catalog No. 23047374 Shop All Microbiologics Products
Change view
Click to view available options
Quantity:
1/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-047-374 1/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-047-374 Supplier Microbiologics™ Supplier No. HE0061S
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Synthetic RNA molecular standard intended for use as a positive control in molecular applications.

  • SARS-CoV-2 synthetic RNA material is a pool containing the following genetic targets:
    • N gene (position 28237-29280) which contains three markers N1, N2 and N3 that are commonly used in diagnostic assays
    • E gene (position 26245-26472)
    • RdRp gene (positions 12641-12824, 13442- 16236)
    • S gene (position 21563-25384)
  • All positions in reference to GenBank Acc. NC_045512.2
  • Dried RNA Preserved with Biomatrica RNAstable™
  • IVD
  • Kit includes:
    • 1 vial of dried synthetic RNA (up to 100 reactions)
    • 1 vial of molecular standard water for rehydration
    • Certificate of Analysis.

Specifications

Biosafety Level 1
Content And Storage 2°C to 25°C
Type Pellet
Packaging Plastic Box
Quantity 1/Pk.
Source United States
Form Pellet
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.