Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zeptometrix SARS-CoV-2 Variant B.1.1.7 (Isolate:USA/CA_CDC_5574/2020) Culture Fluid (Heat Inactivated)
SDP

Catalog No. 22158421 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-158-421 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-158-421 Supplier Zeptometrix Supplier No. 0810612CFHI
Add to Cart
Add to Cart

This product is intended for research, analytical testing during product development, or quality assurance testing. Viral culture fluids are sold as consumable testing materials, and are not for propagation or commercialization.

SARS-Related Coronavirus 2 Lineage B.1.1.7; Alpha Variant (Isolate: USA/CA_CDC_5574/2020) is an enveloped, positive-sense single-stranded RNA virus from the Coronaviridae family and the Betacoronaviridae genus. Severe acute respiratory syndrome-related coronavirus 2 (SARS-CoV-2), isolate USA/CA_CDC_5574/2020 was isolated from a nasopharyngeal swab on December 29, 2020 in San Diego County, California, USA. Under the nomenclature system introduced by GISAID (Global Initiative on Sharing All Influenza Data), SARS-CoV-2, isolate USA/CA_CDC_5574/2020 is assigned lineage B.1.1.7 and GISAID clade GR using Phylogenetic Assignment of Named Global Outbreak LINeages (PANGOLIN) tool. The complete genome of SARS-CoV-2, isolate USA/CA_CDC_5574/2020 has been sequenced (GISAID: EPI_ISL_751801). Each frozen aliquot contains 1 mL of heat inactivated viral culture fluid. The pre-inactivation titer was determined from an infectious aliquot. Viral inactivation is verified after heat inactivation by the absence of viral growth in tissue culture-based infectivity assays.

Specifications

Biosafety Level 2
Certifications/Compliance ISO 13485
Content And Storage Store at -65°C or below
Type Infectious Culture Fluid
For Use With (Application) Nucleic Acid-Molecular Testing, Limit of Detection (LOD) Studies, Cross-reactivity Studies, Other Viral-based Assays
Quantity 1 mL
Regulatory Status RUO

For the purchase of Zeptometrix's BSL2 or BSL3 level products the Laboratories Biosafety Level must be consistent with the products requirement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.