Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zeptometrix SARS-CoV-2 Variant B.1.351 (Isolate: South_Africa/KRISP-K005325/2020) Culture Fluid (Heat Inactivated)
SDP

Catalog No. 22158422 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-158-422 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-158-422 Supplier Zeptometrix Supplier No. 0810613CFHI
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Infectious virus preparation designed to meet research and product development needs.

SARS-Related Coronavirus 2 Lineage B.1.351; Beta Variant (Isolate: South_Africa/KRISP-K005325/2020) is an enveloped, positive-sense single-stranded RNA virus from the Coronaviridae family and the Betacoronaviridae genus. Severe acute respiratory syndrome-related coronavirus 2 (SARS-CoV-2), isolate hCoV-19/South Africa/KRISPK005325/2020 (also referred to as 501Y.V2.HV and 501Y.V2.HV001) was isolated from an oropharyngeal swab from a 40-year-old human male in Ugu district, KwaZulu-Natal, South Africa on November 16, 2020. Under the nomenclature system introduced by GISAID (Global Initiative on Sharing All Influenza Data), SARSCoV-2, isolate hCoV-19/South Africa/KRISPK005325/2020 is assigned lineage B.1.351 and GISAID clade GH using Phylogenetic Assignment of Named Global Outbreak LINeages (PANGOLIN) tool. The complete genome of the clinical isolate of SARS-CoV-2, hCoV-19/South Africa/KRISP-K005325/2020 has been sequenced (GISAID: EPI_ISL_678615). Each frozen aliquot contains 1 mL of heat inactivated viral culture fluid. The pre-inactivation titer was determined from an infectious aliquot. Viral inactivation is verified after heat inactivation by the absence of viral growth in tissue culture-based infectivity assays.

Specifications

Biosafety Level 2
Certifications/Compliance ISO 13485
Content And Storage Store at -65°C or below
Type Infectious Culture Fluid
For Use With (Application) Nucleic Acid-Molecular Testing, Limit of Detection (LOD) Studies, Cross-reactivity Studies, Other Viral-based Assays
Quantity 1 mL
Regulatory Status RUO

For the purchase of Zeptometrix's BSL2 or BSL3 level products the Laboratories Biosafety Level must be consistent with the products requirement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.