Learn More
Cayman Chemical SARS-CoV-2humn nutralIzI 100ug
SDP
Catalog No. 502948052
Encompass_Preferred
Immunogen/Preparation: This antibody was produced as a recombinant protein expressed in CHO cells using engineered constructs based on antibodies from COVID-19 patients. • Host: CHO cells • Species Reactivity: (+) Human • Applications: ELISA, IF, WB
Catalog No. 50-294-8052 Supplier Cayman Chemical Supplier No. 32526100 UG
Immunogen/Preparation: This antibody was produced as a recombinant protein expressed in CHO cells using engineered constructs based on antibodies from COVID-19 patients. • Host: CHO cells • Species Reactivity: (+) Human • Applications: ELISA, IF, WB