Learn More
ACROBiosystems ACROBiosystems SARS-CoV-2 Spike Protein Titer Assay Kit
Catalog No. 502107433
Encompass_Preferred
Kits;96TESTS;This assay kit employs a standard sandwich-ELISA format, providing a rapid detection of SARS-CoV-2 Spike Protein. The kit consists of microplate pre-coated with Anti-SARS-CoV-2 Spike protein Antibody, an SARS-CoV-2 Spike Protein as Control, an biotin-Anti-SARS-CoV-2 Spike protein Antibody, HRP-Streptavidin and buffers.;This assay kit employs a standard sandwich-ELISA format, providing a rapid detection of SARS-CoV-2 Spike protein. The kit consists of pre-coated Anti-SARS-CoV-2 Spike Protein Antibody Microplate, SARS-CoV-2 Spike protein as Control, biotin-Anti-SARS-CoV-2 Spike protein antibody and HRP-Streptavidin. Detection is performed using HRP-Streptavidin with sensitivity of 200 pg/mL.
Catalog No. 50-210-7433 Supplier ACROBiosystems Supplier No. RASA02096TESTS
Kits;96TESTS;This assay kit employs a standard sandwich-ELISA format, providing a rapid detection of SARS-CoV-2 Spike Protein. The kit consists of microplate pre-coated with Anti-SARS-CoV-2 Spike protein Antibody, an SARS-CoV-2 Spike Protein as Control, an biotin-Anti-SARS-CoV-2 Spike protein Antibody, HRP-Streptavidin and buffers.;This assay kit employs a standard sandwich-ELISA format, providing a rapid detection of SARS-CoV-2 Spike protein. The kit consists of pre-coated Anti-SARS-CoV-2 Spike Protein Antibody Microplate, SARS-CoV-2 Spike protein as Control, biotin-Anti-SARS-CoV-2 Spike protein antibody and HRP-Streptavidin. Detection is performed using HRP-Streptavidin with sensitivity of 200 pg/mL.