Learn More
Enzo Life Sciences Coronavirus (SARS-CoV Envelope) antigen (aa 1-76), (recombinant) (100µg)
SDP
Supplier:  Enzo Life Sciences ENZPRT2850100
Encompass_Preferred
Alternative name: Coronavirus envelope protein. Purity: ≥95%. Formulation: Liquid. In 50 mM Tris-HCl, 60 mM NaCl with 50% glycerol. Source: Produced in E. coli. Coronavirus (SARS-CoV) envelope protein (aa 1-76). Long Term Storage: -20°C. Applications: ELISA, WB .
Catalog No. 50-201-4096