Learn More
Enzo Life Sciences Coronavirus (SARS-CoV NP) antigen (aa 340-390), (recombinant) (100µg)
SDP
Supplier:  Enzo Life Sciences ENZPRT2910100
Encompass_Preferred
Alternative name: Coronavirus nucleoprotein. Purity: ≥95%. Formulation: Liquid. In 50 mM Tris-HCl, 60 mM NaCl with 50% glycerol. Source: Produced in E. coli. C-terminal coronavirus (SARS-CoV) nucleoprotein (aa 340-390). Long Term Storage: -20°C. Applications: ELISA, WB .
Catalog No. 50-201-4102