Learn More
Enzo Life Sciences Coronavirus (SARS-CoV NP) antigen, (recombinant) (His-tag) (20µg)
SDP
Supplier:  Enzo Life Sciences ENZPRT2920020
Encompass_Preferred
Alternative name: Coronavirus nucleoprotein. Purity: ≥95%. Formulation: Lyophilized. Source: Produced in E. coli. Coronavirus (SARS-CoV) nucleoprotein (422 aa) is fused to a His-tag. Long Term Storage: -20°C. Applications: ELISA, WB .
Catalog No. 50-201-4103