Learn More
Enzo Life Sciences Coronavirus (SARS-CoV NP) antigen (aa 340-390), (recombinant) (His-tag) (50µg)
SDP
Supplier:  Enzo Life Sciences ENZPRT2950050
Encompass_Preferred
Alternative name: Coronavirus nucleoprotein. Purity: ≥90%. Formulation: Liquid. In 10 mM PBS, pH 7.2. Source: Produced in E. coli. Coronavirus (SARS-CoV) nucleoprotein (aa 340-390) is fused to a His-tag. Long Term Storage: -20°C. Applications: ELISA .
Catalog No. 50-201-4106