Learn More
Medchemexpress LLC SARS-CoV S HEK293 2ug
SDP
Catalog No. 5000196252
Encompass_Preferred
Recombinant SARS-CoV spike protein fragment expressed in HEK293 cells with N-terminal 10xHis and C-terminal Myc tags. The product corresponds to residues R306-F527 (accession P59594) and is provided in small research pack sizes for use in binding studies, antibody characterization, and assay development.
  • Expressed in HEK293 for human-like glycosylation.
  • Contains N-terminal His and C-terminal Myc tags for purification and detection.
  • Corresponds to residues R306-F527 for a defined functional domain.
  • Available in microgram pack sizes suitable for assay optimization.
  • Comes with datasheet, certificate of analysis, and SDS for quality assurance.
Catalog No. 50-001-96252 Supplier Medchemexpress LLC Supplier No. HYP720452UG
Recombinant SARS-CoV spike protein fragment expressed in HEK293 cells with N-terminal 10xHis and C-terminal Myc tags. The product corresponds to residues R306-F527 (accession P59594) and is provided in small research pack sizes for use in binding studies, antibody characterization, and assay development.
  • Expressed in HEK293 for human-like glycosylation.
  • Contains N-terminal His and C-terminal Myc tags for purification and detection.
  • Corresponds to residues R306-F527 for a defined functional domain.
  • Available in microgram pack sizes suitable for assay optimization.
  • Comes with datasheet, certificate of analysis, and SDS for quality assurance.