Learn More
Enzo Life Sciences Coronavirus (SARS-CoV S1) antigen (aa 1-53, 90-115, 171-205), (recombinant) (His-tag) (50µg)
SDP
Supplier:  Enzo Life Sciences ENZPRT2990050
Encompass_Preferred
Alternative name: Coronavirus spike protein. Purity: ≥90%. Formulation: Liquid. In 10 mM PBS, pH 7.2. Source: Produced in E. coli. Coronavirus (SARS-CoV) spike protein (aa 1-53, 90-115, 171-205) is fused to a His-tag. Long Term Storage: -20°C. Applications: ELISA .
Catalog No. 50-201-4110