Learn More
Enzo Life Sciences Coronavirus (SARS-CoV S2) antigen (aa 1051-1076), (recombinant) (100µg)
SDP
Supplier:  Enzo Life Sciences ENZPRT3020100
Encompass_Preferred
Alternative name: Coronavirus spike protein. Purity: ≥95%. Formulation: Liquid. In 25 mM Tris-HCl with detergents and 50% glycerol. Source: Produced in E. coli. C-terminal coronavirus (SARS-CoV) spike protein (aa 1051-1076, 1121-1154, 1162-1190). Long Term Storage: -20°C. Applications: ELISA, WB .
Catalog No. 50-201-4113