Learn More
Enzo Life Sciences Coronavirus (SARS-CoV S2) antigen (aa 408-470, 540-573), (recombinant) (100µg)
SDP
Supplier:  Enzo Life Sciences ENZPRT3010100
Encompass_Preferred
Alternative name: Coronavirus spike protein. Purity: ≥95%. Formulation: Liquid. In 25 mM Tris-HCl with detergents and 50% glycerol. Source: Produced in E. coli. Coronavirus (SARS-CoV) spike protein (aa 408-470, 540-573). Long Term Storage: -20°C. Applications: ELISA, WB .
Catalog No. 50-201-4112