Learn More
Sino Biological SARS-CoV / SARS-CoV-2 Spike Antibody (Biotin), Chimeric MAb 20 µg
Catalog No. 5000704861
Encompass_Preferred
It is a chimeric monoclonal antibody combining the constant domains of the human IgG1 molecule with mouse variable regions. The variable region was obtained from a mouse immunized with purified, recombinant SARS-CoV Spike RBD Protein. The antibody was produced using recombinant antibody technology and then biotinylated.
Catalog No. 50-007-04861 Supplier Sino Biological Supplier No. 40150D003B20
It is a chimeric monoclonal antibody combining the constant domains of the human IgG1 molecule with mouse variable regions. The variable region was obtained from a mouse immunized with purified, recombinant SARS-CoV Spike RBD Protein. The antibody was produced using recombinant antibody technology and then biotinylated.