Learn More
Sino Biological SARS-CoV / SARS-CoV-2 Spike Antibody (HRP), Chimeric MAb 20 µg
Catalog No. 5000704853
Encompass_Preferred
It is a chimeric monoclonal antibody combining the constant domains of the human IgG1 molecule with rabbit variable regions. The variable region was obtained from a rabbit immunized with purified, recombinant SARS-CoV Spike RBD Protein. The antibody was produced using recombinant antibody technology and conjugated with horseradish-peroxidase (HRP).
Catalog No. 50-007-04853 Supplier Sino Biological Supplier No. 40150D006H20
It is a chimeric monoclonal antibody combining the constant domains of the human IgG1 molecule with rabbit variable regions. The variable region was obtained from a rabbit immunized with purified, recombinant SARS-CoV Spike RBD Protein. The antibody was produced using recombinant antibody technology and conjugated with horseradish-peroxidase (HRP).