Learn More
Sino Biological SARS-CoV / SARS-CoV-2 Spike RBD Antibody(HRP), Rabbit PAb, Antigen Affinity Purified 20 µg
Catalog No. 5000749200
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant SARS-CoV Spike/RBD. SARS-CoV Spike RBD specific IgG was purified by SARS-CoV Spike/RBD affinity chromatography and conjugated with horseradish-peroxidase (HRP).
Catalog No. 50-007-49200 Supplier Sino Biological Supplier No. 40150T30H20
Produced in rabbits immunized with purified, recombinant SARS-CoV Spike/RBD. SARS-CoV Spike RBD specific IgG was purified by SARS-CoV Spike/RBD affinity chromatography and conjugated with horseradish-peroxidase (HRP).