Learn More
Enzo Life Sciences Coronavirus (SARS-CoV Spike) polyclonal antibody (100µg)
SDP
Supplier:  Enzo Life Sciences ENZABS8250100
Encompass_Preferred
Isotype: IgG. Immunogen: Peptide corresponding to 17 amino acids near the center of SARS-CoV Spike glycoprotein. The immunogen is located within amino acids 550-600 of SARS-CoV Spike. Species reactivity: Coronavirus. Applications: ELISA. Purity: Affinity purified. Formulation: Liquid. In PBS containing 0.02% sodium azide.
Catalog No. 50-201-3349