Learn More
Sino Biological SARS-CoV Spike S2-His Recombinant Protein 100ug
Supplier:  Sino Biological 40150V08B3100
Encompass_Preferred
A DNA sequence encoding the SARS-CoV spike S2 (NP_828851.1) (Ser668-Pro1195) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-211-5536