Learn More
Invitrogen™ SARS Envelope Protein Synthetic Peptide
Description
SARS Envelope Synthetic Peptide, PEP-0261, from Invitrogen.
Specifications
Specifications
| Species | Virus |
| Purification Method | purified |
| Common Name | SARS Envelope Protein |
| Concentration | 200 μg/mL |
| Content And Storage | -20°C |
| Description | Synthetic Peptide |
| Format | Liquid |
| Formulation | PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 |
| For Use With (Application) | Blocking Assay, Control |
| Lyophilized | No |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.