Learn More
Creative Biomart HBcAg Antigen
Supplier:  Creative Biomart SARS400
Encompass_Preferred
HBcAg subtype adw2 produced in E. coli, is a single, non-glycosylated, polypeptide chain containing 1-149 amino acids and having a molecular weight of approximately 20kDa. HBcAg adw2 1-149 protamine-l
Catalog No. 50-208-4778