Learn More
Sino Biological SARS NP-CoV MAb 20ul (Sino Biological)
Supplier:  Sino Biological 40143R004A20
Encompass_Preferred
It is a chimeric monoclonal antibody combining the constant domains of the human IgG1 molecule with rabbit variable regions, and the variable region was from rabbit monoclonal antibody (Cat#40143-R004). The chimeric monoclonal antibody was produced using recombinant antibody technology.
Catalog No. 50-237-0690