Learn More
Sino Biological SARS NP-CoV PAb 20ul (Sino Biological)
Supplier:  Sino Biological 80715R08H50
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant SARS-CoV Nucleocapsid ( Catalog#40143-V08B; NP_828858.1; Met1-Ala422). Nucleocapsid specific IgG was purified by SARS-CoV Nucleocapsid affinity chromatography.
Catalog No. 50-237-4185