Learn More
Creative Biomart Recombinant SARS Nucleocaspid Protein, GST-tagged
Supplier:  Creative Biomart SARS403
Encompass_Preferred
The E. coli derived 32 kDa recombinant protein contains the Nucleocapsid core protein 1-49 amino acids immunodominant regions. SARS Necleocapsid protein 1-49 a.a. is fused to a GST tag.
Catalog No. 50-208-4781