Learn More
LifeSensors SARS NUCLEOCAPSID PROTEIN ANTI
SDP
Supplier:  LifeSensors CV40010100
Encompass_Preferred
Within the last two decades, SARS and MERS coronaviruses emerged as global health concerns causing severe acute respiratory syndromes. In December 2019, a novel coronavirus (SARS-CoV-2) was identified in Wuhan, Hubei province in China. The SARS-CoV genome encodes several structural proteins including the nucleocapsid protein (Nucleoprotein; Protein N), which plays a role in virion assembly through its interaction with the viral genome and the membrane protein. This key protein packages the positive strand viral RNA into a helical ribonucleocapsid. The nucleocapsid is a highly conserved and immunogenic viral protein thus representing a valuable tool for diagnostic and vaccine production purposes. The SARS Nucleocapsid Protein Antibody recognizes and binds to both SARS-CoV and SARS-CoV-2 nucleocapsid proteins.
Catalog No. NC1836133