Learn More
Creative Biomart Recombinant SARS Nucleocaspid Protein, His-tagged
Supplier:  Creative Biomart SARS406
Encompass_Preferred
The HEK293 derived recombinant protein contains the SARS Coronavirus Nucleoprotein, amino acids 2-422 fused to His tag at C-terminal.
Catalog No. 50-208-4784