Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix SARS-Related Coronavirus 2 (SARS-CoV-2) External Run Control
SDP

Catalog No. 22158370 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
6 x 0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-158-370 6 x 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-158-370 Supplier ZeptoMetrix Supplier No. NATSARS(COV2)ERC
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SARS-Related Coronavirus 2 (SARS-CoV-2) External Run Control is formulated with purified, intact viral particles (Positive control) and human A549 cells (Negative control).

  • The virus particles have been chemically modified to render them non-infectious and refrigerator stable.
  • These controls are provided in a proprietary matrix.
  • NATtrol™ SARS-CoV-2 external run controls are designed to evaluate the performance of nucleic acid tests for determination of the presence of SARSCoV-2 RNA. NATSARS(COV2)-ERC and NATSARS(COV2)-NEG enable laboratories to monitor test variation, lot-to-lot test kit performance, operator variation, and can provide assistance in identifying random or systemic error.
  • Extract nucleic acid prior to use in downstream assays that are not sample to result.
  • Quality control materials should be used in accordance with local, state, federal, and accreditation requirements.

Specifications

Content And Storage Store at 2°C to 8°C
Description Mix tube vigorously for at least 5 seconds
Concentration 50,000 copies/mL
For Use With (Equipment) To avoid cross-contamination, use separate pipette tips for all materials
Type SARS-CoV-2 (Isolate: USA-WA1/2020)
Includes Each control pack contains 6 x 0.5 mL of NATtrol™ SARS-CoV-2 or A549 cells
Quantity 6 x 0.5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.