Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix SARS-Related Coronavirus 2 (SARS-CoV-2) Stock
SDP

Catalog No. 22158371 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
1 mL/Vial
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-158-371 1 mL/Vial
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-158-371 Supplier ZeptoMetrix Supplier No. NATSARS(COV2)ST
 Restricted Item
Add to Cart
Add to Cart

SARS-Related Coronavirus 2 (SARS-CoV-2) Stock is formulated with purified, intact viral particles.

  • The virus particles have been chemically modified to render them non-infectious and refrigerator stable.
  • NATtrol™ SARS-CoV-2 stock is designed to evaluate the performance of nucleic acid tests for determination of the presence of SARS-CoV-2 RNA. NATtrol™ SARS-CoV-2 stock enables laboratories to monitor test variation, lot-to-lot test kit performance, operator variation, and can provide assistance in identifying random or systemic error.
  • Extract nucleic acid prior to use in downstream assays that are not sample to result.
  • Quality control materials should be used in accordance with local, state, federal, and accreditation requirements.

Specifications

Content And Storage Store at 2°C to 8°C
Description Mix tube vigorously for at least 5 seconds
Concentration 1,000,000 copies/mL
Type Stock
Includes Each vial contains 1.0 mL of NATtrol™ SARS-CoV-2 stock in a proprietary matrix
Quantity 1 mL/Vial
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.