Learn More
Creative Biomart Recombinant SARS Spike Protein, His-tagged
Supplier:  Creative Biomart SARS409
Encompass_Preferred
The HEK293 derived recombinant protein contains the SARS Coronavirus Spike S1 Gycoprotein, amino acids 1-666 fused to His tag at C-terminal.
Catalog No. 50-208-4787