Learn More
Sino Biological SARS Spike MAb 20ul (Sino Biological)
Supplier:  Sino Biological 40150MM1020
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant SARS-CoV Spike S1 Subunit ( Catalog#40150-V08B1; AAX16192.1; Met1-Arg667). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-3774