Learn More
Sino Biological SARS Spike MAb 20ul (Sino Biological)
Supplier:  Sino Biological 40150D00620
Encompass_Preferred
It is a chimeric monoclonal antibody combining the constant domains of the human IgG1 molecule with rabbit variable regions. The variable region was obtained from a rabbit immunized with purified, recombinant SARS-CoV Spike RBD Protein. The antibody was produced using recombinant antibody technology.
Catalog No. 50-237-2032