Learn More
Medchemexpress LLC Sarsasapogenin | 126-19-2 | 416.64 | 50 MG
SDP
Catalog No. 5000427894
Encompass_Preferred
Sarsasapogenin is a sapogenin from the Chinese medical herb *Anemarrhena asphodeloides Bunge*, with antidiabetic, anti-oxidative, anticancer, and anti-inflammatory activities.
  • Derived from *Anemarrhena asphodeloides Bunge*, a Chinese medical herb.
  • Exhibits antidiabetic activity.
  • Possesses anti-oxidative properties.
  • Demonstrates anticancer activity.
  • Has anti-inflammatory activities.
Catalog No. 50-004-27894 Supplier Medchemexpress LLC Supplier No. HYN0073R50MG
Sarsasapogenin is a sapogenin from the Chinese medical herb *Anemarrhena asphodeloides Bunge*, with antidiabetic, anti-oxidative, anticancer, and anti-inflammatory activities.
  • Derived from *Anemarrhena asphodeloides Bunge*, a Chinese medical herb.
  • Exhibits antidiabetic activity.
  • Possesses anti-oxidative properties.
  • Demonstrates anticancer activity.
  • Has anti-inflammatory activities.