Learn More
Medchemexpress LLC Sarsasapogenin | 126-19-2 | MFCD00270414 | 99.9% | 416.64 | C27H44O3 | 10 MG
SDP
Catalog No. 5000296993
Encompass_Preferred
Sarsasapogenin is a steroidal sapogenin supplied as a high-purity analytical standard for research and analytical applications. It is provided as a white to off-white solid with documented specifications used for reference and analytical work.
  • High purity reference standard (99.9%).
  • Molecular formula C27H44O3 and molecular weight 416.64.
  • Supplied as a 10 mg solid, white to off-white.
  • Intended for analytical and research applications as a calibration or reference material.
  • Store under conditions specified in the certificate of analysis.
Catalog No. 50-002-96993 Supplier Medchemexpress LLC Supplier No. HYN0073R10MG
Sarsasapogenin is a steroidal sapogenin supplied as a high-purity analytical standard for research and analytical applications. It is provided as a white to off-white solid with documented specifications used for reference and analytical work.
  • High purity reference standard (99.9%).
  • Molecular formula C27H44O3 and molecular weight 416.64.
  • Supplied as a 10 mg solid, white to off-white.
  • Intended for analytical and research applications as a calibration or reference material.
  • Store under conditions specified in the certificate of analysis.