Learn More
Medchemexpress LLC HY-N0073 25mg Medchemexpress, Sarsasapogenin CAS:126-19-2 Purity:>98%
SDP
Catalog No. 501933379
Encompass_Preferred
Medchemexpress, HY-N0073 25mg Sarsasapogenin CAS:126-19-2 Sarsasapogenin is a sapogenin from the Chinese medical herb Anemarrhena asphodeloides Bunge, with antidiabetic, anti-oxidative, anticancer and anti-inflamatory activities. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3379 Supplier Medchemexpress LLC Supplier No. HYN007325MG
Medchemexpress, HY-N0073 25mg Sarsasapogenin CAS:126-19-2 Sarsasapogenin is a sapogenin from the Chinese medical herb Anemarrhena asphodeloides Bunge, with antidiabetic, anti-oxidative, anticancer and anti-inflamatory activities. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.