Learn More
Medchemexpress LLC Sarsasapogenin (Standard) | 126-19-2 | 99.91% | 416.64 | 5 MG
SDP
Catalog No. 5000427992
Encompass_Preferred
Sarsasapogenin (Standard) is the analytical standard of Sarsasapogenin, intended for research and analytical applications. Sarsasapogenin is a sapogenin found in the Chinese medical herb Anemarrhena asphodeloides Bunge, possessing antidiabetic, anti-oxidative, anticancer, and anti-inflammatory activities.
  • Used as a reference standard for assays
  • Utilized in qualitative, quantitative, and methodological research experiments
  • Suitable for HPLC, GC, and MS applications
Catalog No. 50-004-27992 Supplier Medchemexpress LLC Supplier No. HYN0073R5MG
Sarsasapogenin (Standard) is the analytical standard of Sarsasapogenin, intended for research and analytical applications. Sarsasapogenin is a sapogenin found in the Chinese medical herb Anemarrhena asphodeloides Bunge, possessing antidiabetic, anti-oxidative, anticancer, and anti-inflammatory activities.
  • Used as a reference standard for assays
  • Utilized in qualitative, quantitative, and methodological research experiments
  • Suitable for HPLC, GC, and MS applications