Learn More
Sino Biological SARS-CoV (Isolate Tor2) Spike RBD Protein (His & AVI Tag), Biotinylated
Catalog No. 502246769
Encompass_Preferred
A DNA sequence encoding the SARS-CoV (isolate Tor2) spike RBD (NP_828851.1) (Arg306-Phe527) was expressed with a c-terminal AVI tagged polyhistidine tag at the C-terminus. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-224-6769 Supplier Sino Biological Supplier No. 40634V27HB20
A DNA sequence encoding the SARS-CoV (isolate Tor2) spike RBD (NP_828851.1) (Arg306-Phe527) was expressed with a c-terminal AVI tagged polyhistidine tag at the C-terminus. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.