Learn More
Cayman Chemical SARSCoV NucleocapsId ProteIn
SDP
Catalog No. 502992089
Encompass_Preferred
Immunogen Fusion protein from the cytoplasmic loop of the Y2 subunit of the rat GABAA receptor Host Rabbit Species Reactivity ( ) Mouse rat Applications IHC WB MW 46 kDa
Catalog No. 50-299-2089 Supplier Cayman Chemical Supplier No. 31430100 UL
Immunogen Fusion protein from the cytoplasmic loop of the Y2 subunit of the rat GABAA receptor Host Rabbit Species Reactivity ( ) Mouse rat Applications IHC WB MW 46 kDa