Learn More
Sino Biological SARS-CoV / SARS-CoV-2 Nucleocapsid Antibody (HRP), Rabbit MAb 100ug
Supplier:  Sino Biological 40143R004H100
Encompass_Preferred
This product is a recombinant monoclonal antibody expressed from HEK293 cells and conjugated with horseradish-peroxidase (HRP).
Catalog No. 50-224-7377