Learn More
Sino Biological SARS-CoV / SARS-CoV-2 Nucleocapsid Antibody (Biotin), Mouse MAb
Supplier:  Sino Biological 40143MM05B100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant SARS-CoV Nucleoprotein / NP (Catalog#40143-V08B; NP_828858.1; Met1-Ala422) and then biotinylated. The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography and then biotinylated.
Catalog No. 50-228-2336