Learn More
Sino Biological SARS-CoV / SARS-CoV-2 Nucleocapsid Antibody(HRP), Mouse MAb 100ug
Supplier:  Sino Biological 40143MM05H100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant SARS-CoV Nucleoprotein / NP (Catalog#40143-V08B; NP_828858.1; Met1-Ala422). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography and conjugated with horseradish-peroxidase (HRP).
Catalog No. 50-224-6464