Learn More
Sino Biological SARS-CoV / SARS-CoV-2 Spike Antibody (Biotin), Chimeric MAb 100ug
Supplier:  Sino Biological 40150D001B100
Encompass_Preferred
It is a chimeric monoclonal antibody combining the constant domains of the human IgG1 molecule with mouse variable regions. The variable region was obtained from a mouse immunized with purified, recombinant SARS-CoV Spike RBD Protein. The antibody was produced using recombinant antibody technology and then biotinylated.
Catalog No. 50-224-6465