Learn More
Sino Biological SARS-CoV-2 (2019-nCoV) Nucleocapsid Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 40588T30100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant SARS-CoV-2 / 2019-nCoV Nucleocapsid Protein (Catalog#40588-V08B; YP_009724397.2(335Gly/Ala); Met1-Ala419). The specific IgG was purified by SARS-CoV-2 / 2019-nCoV Nucleocapsid affinity chromatography.
Catalog No. 50-224-7775