Learn More
Sino Biological SARS-CoV-2 (2019-nCoV) Spike RBD Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Catalog No. 502247480
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant SARS-CoV-2 (2019-nCoV) Spike RBD (YP_009724390.1; Arg319-Phe541). SARS-CoV-2 (2019-nCoV) Spike RBD specific IgG was purified by SARS-CoV-2 (2019-nCoV) Spike RBD affinity chromatography.
This item is not returnable. View return policy
Catalog No. 50-224-7480 Supplier Sino Biological Supplier No. 40592T30100
Produced in rabbits immunized with purified, recombinant SARS-CoV-2 (2019-nCoV) Spike RBD (YP_009724390.1; Arg319-Phe541). SARS-CoV-2 (2019-nCoV) Spike RBD specific IgG was purified by SARS-CoV-2 (2019-nCoV) Spike RBD affinity chromatography.