Learn More
Sino Biological SARS-CoV-2 (2019-nCoV) Spike S1+S2 (L18F, T20N, P26S, D138Y, R190S, K417T, E484K, N501Y, D614G, H655Y, T1027I, V1176F) Protein (ECD, His Tag)
Supplier:  Sino Biological 40589V08B10100
Encompass_Preferred
A DNA sequence encoding the SARS-CoV-2 (2019-nCoV) Spike S1+S2 (YP_009724390.1, with mutations L18F, T20N, P26S, D138Y, R190S, K417T, E484K, N501Y, D614G, H655Y, T1027I, V1176F) (Met1-Pro1213) was expressed with a polyhistidine tag at the C-terminus. The mutations were identified in the SARS-CoV-2 variant (known as variant P.1) which emerged in the Brazil.
Catalog No. 50-211-4274