Learn More
Cayman Chemical SARSCoV2 B 1 1 7 SpIke Reag
SDP
Supplier:  Cayman Chemical 502181100 DTN
Encompass_Preferred
SARS-CoV-2 B.1.1.7 Spike Reagent has been tested and formulated to work exclusively with Cayman’s SARS-CoV-2 (B.1.1.7, Alpha Variant) Neutralizing Antibody SimpleDetect ELISA Kit (Item No. 502180).
Catalog No. 50-289-2243