Learn More
Sino Biological SARS-CoV-2 (B.1.1.529 sublineage BA.2) Spike S1 Protein (His Tag) (HPLC-verified)
Supplier:  Sino Biological 40591V08H43100
Encompass_Preferred
A DNA sequence encoding the SARS-CoV-2 Spike S1 (YP_009724390.1, with mutation T19I, L24S, del25-27, G142D, V213G, G339D, S371F, S373P, S375F, T376A, D405N, R408S, K417N, N440K, S477N, T478K, E484A, Q493R, Q498R, N501Y, Y505H, D614G, H655Y, N679K, P681H) (Met1-Arg685) was expressed with a polyhistidine tag at the C-terminus. The mutations were identified in the SARS-CoV-2 variant (known as variant B.1.1.529 sublineage BA.2).
Catalog No. 50-224-6489