Learn More
Sino Biological SARS-CoV-2 (B.1.1.7 + E484K) Spike S1+S2 trimer Protein (ECD, His tag) (HPLC-verified)
Supplier:  Sino Biological 40589V08H17100
Encompass_Preferred
A DNA sequence encoding the SARS-CoV-2 Spike S1+S2 (YP_009724390.1, with mutations T19R, E156G, DEL157-158, L452R, T478K, D614G, P681R, D950N,F817P, A892P, A899P,A942P, K986P, V987P and furin cleavage site mutants) expressed with the bacteriophage T4 fibritin and a polyhistidine tag at the C-terminus. The mutations were identified in the SARS-CoV-2 variant (known as variant B.1.1.7 +E484K).
Catalog No. 50-224-6476