Learn More
Sino Biological SARS-CoV-2 B.1.351 Spike S1+S2 trimer Protein (ECD, His tag) 100ug
Supplier:  Sino Biological 40589V08H13100
Encompass_Preferred
A DNA sequence encoding the SARS-CoV-2 Spike S1+S2 (YP_009724390.1, with mutations L18F, D80A, D215G, 241-243 deletion, R246I, K417N, E484K, N501Y, D614G, A701V, F817P, A892P, A899P, A942P, K986P, V987P and furin cleavage site mutants) expressed with the bacteriophage T4 fibritin and a polyhistidine tag at the C-terminus. The mutations were identified in the SARS-CoV-2 variant (known as variant B.1.351) which emerged in the South Africa.
Catalog No. 50-224-6623