Learn More
Sino Biological SARS-CoV-2 (B.1.640.2) Spike RBD Protein (His Tag) (HPLC-verified)
Supplier:  Sino Biological 40592V08H128100
Encompass_Preferred
A DNA sequence encoding the SARS-CoV-2 Spike RBD (YP_009724390.1, with mutation R346S, N394S, Y449N, E484K, F490R, N501Y) (Arg319-Phe541) was expressed with a polyhistidine tag at the C-terminus. The mutations were identified in the SARS-CoV-2 variant (known as variant B.1.640.2).
Catalog No. 50-224-6496