Learn More
Sino Biological SARS-CoV-2 (BA.4/BA.5/BA.5.2) Spike S1+S2 trimer Protein (ECD, His & AVI Tag), Biotinylated (HPLC-verified)
Catalog No. 502282130
Encompass_Preferred
A DNA sequence encoding the SARS-CoV-2 (BA.4/BA.5/BA.5.2) Spike S1+S2 trimer (YP_009724390.1, with mutations T19I, L24del, P25del, P26del, A27S, H69del, V70del, G142D, V213G, G339D, S371F, S373P, S375F, T376A, D405N, R408S, K417N, N440K, L452R, S477N, T478K, E484A, F486V, Q498R, N501Y, Y505H, D614G, H655Y, N679K, P681H, N764K, D796Y, F817P, A892P, A899P, A942P, Q954H, N969K, K986P, V987P and furin cleavage site mutants) (Met1-1208Q) was expressed with bacteriophage T4 fibritin and a C-terminal polyhistidine tag followed by an AVI tag. The mutations were identified in the SARS-CoV-2 variant (known as variant BA.4/BA.5/BA.5.2).The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
This item is not returnable. View return policy
Catalog No. 50-228-2130 Supplier Sino Biological Supplier No. 40589V49H5B20
A DNA sequence encoding the SARS-CoV-2 (BA.4/BA.5/BA.5.2) Spike S1+S2 trimer (YP_009724390.1, with mutations T19I, L24del, P25del, P26del, A27S, H69del, V70del, G142D, V213G, G339D, S371F, S373P, S375F, T376A, D405N, R408S, K417N, N440K, L452R, S477N, T478K, E484A, F486V, Q498R, N501Y, Y505H, D614G, H655Y, N679K, P681H, N764K, D796Y, F817P, A892P, A899P, A942P, Q954H, N969K, K986P, V987P and furin cleavage site mutants) (Met1-1208Q) was expressed with bacteriophage T4 fibritin and a C-terminal polyhistidine tag followed by an AVI tag. The mutations were identified in the SARS-CoV-2 variant (known as variant BA.4/BA.5/BA.5.2).The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.